Spécifications
- Description:Soft-Sided Field Case
- Cat. no.:10010-474
- Supplier No.:1210005
Spécifications
A propos de cet article
This cushioned field case with interior dividers and exterior pockets protects meters, electrodes, solution, and beakers under field conditions.
Designed for use with Orion™ Star™ A120 series portable meters; Orion™ Star™ A220 series portable meters; Orion™ Star™ A320 series portable meters; Orion™ 3-Star, 4-Star and 5-Star series portable meters; Orion™ Star™ and Star Plus portable meters.