Spécifications
- Description:Protective Meter Armor
- Cat. no.:89206-432
- Supplier No.:STARA-AR
Spécifications
A propos de cet article
This portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder, and pH electrode storage sleeve.
For use with Orion™ Star™ A series portable meters (Star A320, Star A220 and Star A120 series).