Spécifications
- Description:Hard-Sided Field Case
- Cat. no.:89206-424
- Supplier No.:STARA-CS
Spécifications
A propos de cet article
This hard-sided field case can be used to protect, transport, and store measuring equipment in the field.
The field case holds Star™ A series portable meters (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe, and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case, and the internal separating door can be removed for cable management.
Designed for use with Orion™ Star™ A series portable meters (Star A320, Star A220 and Star A120 series).