A propos de cet article
Manual syringes for use with Beckman/Altex, Rheodyne, SSI, and Valco Valves.
- 5 µL to 500 µL syringe volumes
When using the complete loop fill technique, the syringe capacity should be greater than twice the loop volume
For the partial loop fill technique, the injection volume should be no greater than half the loop capacity. The injection size sets the injection volume.
Note: Users of older Valco injectors requiring 3/4" long needles must fit a Valco VISF-2 adapter.
The loop capacity sets the injection volume.