Orion™ Star™ A Series Electrode Holders for Portable Meters, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Electrode holders can be used with the Star™ A series portable meter armor and storage sleeve to protect, transport, and store sensors in the field.
- Can be attached to either side of the portable meter armor
- Holder for pH electrodes holds one pH electrode when placed in the storage sleeve
- Holder for conductivity and DO probes holds 12mm or 15mm diameter conductivity probes and a dissolved oxygen probe when placed in calibration sleeve
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.