Certified Reference Materials, Sulphide, KLEN INTERNATIONAL
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Certificates
About this item
KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels.
Ordering information: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.
Frequently Bought Together