Ammonium sulfate, BiotechGrade
Fournisseur: Spectrum Chemicals
Certificats
A propos de cet article
Synonymes: Sulphuric acid diammonium salt
Formule:(NH₄)₂SO₄
MW:132.14 g/mol
Numéro MDL:MFCD00003391
Numéro CAS:7783-20-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 336 à 339 °C
:Pt Eclair:|Point eclaire:
:M.W. 132,14 g/mol
:GHS07
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 336 to 339 °C
:Flash Pt:
:Flash Pt:
:M.W. 132.14 g/mol
Température d'entreposage:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Résultats des tests de spécification
- Appearance:White, Crystalli
- Assay [(NH₄)₂SO₄]:99.0 % min.
- pH of a 5% Solution @ 25°C:5.0 - 6.0
- Solubility (10G in 50ML):Clear, Colorless
- Insoluble Matter:max. 0.005 %
Résultats des tests de spécification
Customers Who Bought This Also Bought