Flask Brush, Justman Brush
# de catalogue 17156-194
Fournisseur: Justman Brush
Spécifications
- Brush Head Diameter:38 mm (1 1/2")
- Handle Material:Wire
- Description:Flask Brush
- Total length:30.5 cm (12")
- Cat. no.:17156-194
- Supplier No.:1781-1
- Brush Head Length:4.25" (10.8 cm)
Spécifications
A propos de cet article
Brush for cleaning various types of flasks.
Customers Who Bought This Also Bought