Flow Meter, Nitrogen, with Metering Valve, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Precision machined clear acrylic Nitrogen flowmeter with brass metering valve and Buna-N seals for use with Chemglass process reactors.
- Front of flowmeter has a blue scale indicating the L/min. of air
- HDPE hose connection is for use with 5/16” ID tubing
Delivery: Unit is supplied complete with a 1/2" O.D. mounting arm for ease in clamping to support frame. Flowmeters with larger L/min. ranges are available.
Spécifications
- Catalog No:
- 16002-788
- 89028-648
- 16002-790
- Body material:
- Clear acrylic
- Clear acrylic
- Clear acrylic
- Max. operating temperature:
- 65°C (150°F)
- 65°C (150°F)
- 65°C (150°F)
- Max. pressure:
- 100.00 psi (6.89 Bar)
- 100.00 psi (6.89 Bar)
- 100.00 psi (6.89 Bar)
- Port:
- 8 mm (Hose Barb O.D.)
- 8 mm (Hose Barb O.D.)
- 8 mm (Hose Barb O.D.)
- Width:
- 25.4 mm
- 25.4 mm
- 25.4 mm
- Height:
- 102 mm
- 102 mm
- 102 mm
- Depth:
- 28.6 mm
- 28.6 mm
- 28.6 mm