Schwartz Drying Tubes, Chemglass
Catalog # 80068-002
Supplier: Chemglass
Small Business Enterprise
Specifications
- ST Joint No.:14/20
- Description:Schwartz Drying Tubes
- Cat. no.:80068-002
- Tube Length:170 mm
- Supplier No.:CG-1297-01
Specifications
About this item
Schwartz type drying tube having hollow stoppers.
Stoppers have a hole that aligns with a serrated hose connection for on/off action.
Customers Who Bought This Also Bought