Chemware® Tongs, Saint-Gobain Performance Plastics
# de catalogue 62448-106
Fournisseur: Saint Gobain Performance Plastics
Spécifications
- Length:24.1 cm (91/2")
- Description:PTFE Fluoropolymer Coated Tongs
- Cat. no.:62448-106
- Supplier No.:D1069290
Spécifications
A propos de cet article
Chemware® PTFE Fluoropolymer Coated Tongs are durable stainless steel tongs covered with an extra-heavy, non-stick PTFE coating that will not scratch or damage delicate surfaces.
- Coated tension screw will not corrode
- Opposing grip handle with contoured jaw for easy operation
- Tapered tips for enhanced grip
Tongs are resistant to high and low temperatures and able to withstand the most severe laboratory conditions.
Customers Who Bought This Also Bought