ContactUsLinkComponent
Plastic Box with Lid, Althor Products
Plastic Box with Lid, Althor Products
Catalog # 470151-928
Plastic Box with Lid, Althor Products
Catalog # 470151-928
Specifications
  • Description:
    Plastic Storage Box
  • Dimensions:
    4×4×2" (W×L×H)
  • Cat. no.:
    470151-928
  • Supplier No.:
    SM1212
Specifications
About this item
Crystal clear tall plastic boxes are supplied with lids that are 1/4" in depth.
  • Easy access to contents
  • Fitted tops
  • Snug edged corners
Customers Who Bought This Also Bought