TRIonic® Tri-Polymer Gloves, Mapa Professional AdvanTech
Fournisseur: MAPA
Certificats
A propos de cet article
Unique tri-polymer construction offers 100% comfort for optimal mechanical and chemical resistance. Tri-polymer blend of neoprene, nitrile, and natural rubber combines comfort with added strength, tear resistance, and chemical splash protection.
- Low level of contaminants
- Extensive range for all applications
- Wide cuff for double-gloving
- Extended length to protect the forearm
This product is not classified as hazardous according to the directive 1999/45/EC of the European Parliament and of the Council.
This product does not contain any substances of very high concern according to the regulation n° 1907/2006 of the European Parliament and of the Council (REACH).
Certifications: Compatible with ISO 4 class 10.
Spécifications
- Catalog No:
- 32916-354
- Ambidextrous/Hand-specific:
- Hand-specific