Spécifications
- Capacity:60 ml (2 oz.)
- Color:Translucent
- Description:Medicine Cups
- Cat. no.:89219-006
- Supplier No.:02102A
Spécifications
A propos de cet article
This medicine cups are used in health care, dental care and other human service industries is to provide a safe way to distribute medication to patients
- Non sterile
- Easy to read gradations
A convenient way to distribute medicine in pill or liquid format.