PTFE-Faced Silicone Rubber Septa, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Aluminium seal with Medium durometer PTFE/silicone septum.
- Good resealability
- Core resistance
- Multiple injections and easy penetration
Customers Who Bought This Also Bought