ContactUsLinkComponent
PIG® Grippy® Plunger Mat, New Pig
PIG® Grippy® Plunger Mat, New Pig
Catalog # 75982-604
Supplier:  New Pig
undefined
PIG® Grippy® Plunger Mat, New Pig
Catalog # 75982-604
Supplier:  New Pig
Supplier Number:  GRP7005-BK
Specifications
  • Description:
    PIG® Grippy® Plunger Mat
  • Length:
    23 cm (9")
  • Cat. no.:
    75982-604
  • Width:
    23 cm (9")
Specifications
About this item
Control messes from toilet plungers and brushes with adhesive-backed PIG® Grippy® Plunger Mats for a more professional restroom environment.
  • Place underneath toilet plungers, brushes, soap dispensers, and other small-footprint areas in the restroom to control drips and eliminate puddles
  • Proprietary adhesive formulation applied across entire backing of mats ensures they stay in place with no shifting, curling, or flipping up
  • Permanently infused with antimicrobial technology to prevent the growth of odor-causing bacteria and mold
  • Top surface is engineered to absorb urine and water on contact, wick it away, and dry out quickly
  • Durable, low-profile mats can be mopped without shifting or sliding
  • Two impermeable backing layers ensure zero liquid pass-through, protecting floorsand grout against degradation and discoloration from uric acid