Topaz GC Inlet Liners for Shimadzu Gas Chromatographs, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
- Topaz technology deactivation
- Low breakdown
- Reproducible
- Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Customers Who Bought This Also Bought