Topaz GC Inlet Liners for PerkinElmer Gas Chromatographs, Packed, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
- Topaz technology deactivation
- Low breakdown
- Reproducible
- Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Certifications: Patented.
Customers Who Bought This Also Bought