Aluminium Heating Blocks, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Ideal for use in place of a sand bath in microscale experiments.
- Excellent heat transfer
- Does not interfere with magnetic stirring
Customers Who Bought This Also Bought