19 - Series Steel Frame Cabinet, Akro-Mils
Fournisseur: Akro-Mils
Certificats
A propos de cet article
19-series cabinets are wall-mountable small parts storage cabinets available in a variety of drawer sizes and drawer colors.
- Chip-proof, durable powder-coated finish
- Heavy-duty 24-gauge, welded-steel frame for strength
- Built-in nesting feet for safe, stable stacking
- Label holder and transparent window for content identification
- Made in the USA
Steel cabinets stack together for highdensity storage. Color drawer options create quick visual reference for easy access to parts.
Customers Who Bought This Also Bought