Linear Diffraction Glasses
# de catalogue 470225-846
Fournisseur: RAINBOW SYMPHONY INC SE
Spécifications
- Description:Diffraction Glasses, Linear, 500 lines/mm
- Cat. no.:470225-846
- Supplier No.:01616
Spécifications
A propos de cet article
Make the study of light and color fun and easy
- Uses a linear diffraction grating
- Easily see identifiable spectral images
Highly efficient, 500 lines per millimeter, single axis holographic diffraction grating lenses separate light from any source into its spectral components for study and analysis. Recommended for use with Power Supply and Spectrum Gas Tubes.
Customers Who Bought This Also Bought