Lab Jacks, Boekel Industries
Fournisseur: Boekel
Certificats
A propos de cet article
These lab jacks provide stable height adjustment for various items in the labs such as hotplates, baths, and small equipment.
- Top and bottom plates are constructed of anodized aluminum
- Internal supports and drive screw are constructed of stainless steel
- Three sizes have a pre-tapped hole to accommodate an optional support rod
- Giant size has a side locking knob for the platform
Caution: Not for lifting loads.
Customers Who Bought This Also Bought