White Urea Closures with PTFE-Faced White Rubber Liners, Kimble Chase, DWK Life Sciences
Catalog # 66009-258
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap size:22-400
- Color:White
- Cat. no.:66009-258
- Supplier No.:74526-22400
Specifications
About this item
Closure is made from white urea and features a foam-backed F-217 PTFE liner which resists attack from virtually all chemicals at room temperature.
- Top is suitable for marking
- Ideal for scintillation vials
For long term sample storage with resilience for a tight seal.
Customers Who Bought This Also Bought