Wave Demonstration Kit
# de catalogue 470225-336
Fournisseur: American Scientific
Spécifications
- Description:Wave Demonstration Kit
- Cat. no.:470225-336
- Supplier No.:8430-01-10
Spécifications
A propos de cet article
Compact kit for all your wave demonstration needs.
- Designed to go along with a mechanical wave driver
This wave demonstration kit includes an elastic cord, resonance hoop and strips, a spring and an attachment hook.
Customers Who Bought This Also Bought