ContactUsLinkComponent
Component Holder
Component Holder
Catalog # 470225-196
Supplier:  VWR International
HOLDER COMPONENT
Component Holder
Catalog # 470225-196
Supplier:  VWR International
Specifications
  • Description:
    Component Holder
  • Cat. no.:
    470225-196
  • Supplier No.:
    PH60106
Specifications
About this item
Makes hard to connect components easy to use in electrical circuits.
  • With 4 mm socket connectors
The component holder is an instrument for holding the electronic components such as resistors, capacitors, LEDs, LDRs etc for the use in different experimental circuits.
Customers Who Bought This Also Bought