Extension Tubing, PTFE, for Bottle Top Dispensers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Autoclavable PTFE delivery Jet-Pen and extension tubing aids dispensing into vessels with maximum comfort within a 60cm distance.
Customers Who Bought This Also Bought