Dabigatran Etexilate Mesylate (DEM) Oversize 24.5 mm ID Baskets and Basket Shafts, Quality Lab Accessories
Supplier: QLA
Certificates
About this item
QLA manufactures Oversize 24.5 mm ID Baskets and Basket Shafts for most dissolution baths.
- Made of high quality 316 stainless steel
- All baskets and shafts are made in the USA
- All baskets and shafts are laser marked with a unique serial number
- 40 Mesh baskets are made from sintered mesh
- Custom length shafts and other mesh sizes are available upon request
Dabigatran Etexilate Mesylate (DEM) requires the use of a special wider basket for dissolution testing. This oversize clip style basket is larger than the standard USP Apparatus 1 basket and will not fit on to a standard basket hub. A special clip style hub/basket shaft must be used.
Customers Who Bought This Also Bought