Dissolution Basket Shafts, Apparatus 1, Quality Lab Accessories
Fournisseur: QLA
Certificats
A propos de cet article
QLA offers both Clip Style and O-ring Push-on style Basket Shafts for most dissolution baths.
- Made of high quality 316 Electropolished Stainless Steel
- All basket shafts made in the USA
- All basket shafts are laser marked with a unique serial number
- Spring clips on clip style basket shafts
- Other shaft lengths are available
Clip style shafts utilize a proprietary, spring tempered steel clip, which retains the original gripping tension far longer than conventional shafts and comply with USP (711) and Original Equipment Manufacturer's dimensional specifications. O-ring push-on style shafts comply with Original Equipment Manufacturer's dimensional specifications and are guaranteed to properly interface with the designated dissolution basket. Both clip on style and O-ring style shafts are precision manufactured to limit rotational wobble to levels below USP (711) specifications.
Certifications: All basket shafts are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an additional cost.
Customers Who Bought This Also Bought