ContactUsLinkComponent
sPLA₂ Assay Kit, Cayman Chemical
sPLA₂ Assay Kit, Cayman Chemical
# de catalogue 75817-226
Numéro CAS:  
KIT SPLA2 ASSAY 405-414 NM 96 WELLS
sPLA₂ Assay Kit, Cayman Chemical
# de catalogue 75817-226
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • No. of wells:
    96
  • Cat. no.:
    75817-226
  • Includes:
    Kit contains: Substrate, Control, Buffer, Reagents, and 96-Well Plate
  • Supplier No.:
    765001-96
Spécifications
A propos de cet article
For the measurement of sPLA2 activity.
  • Measure sPLA2 activity
  • Assay 46 samples in duplicate
  • Measure sPLA2 activity down to 0.02 U/ml
  • Plate-based colorimetric measurement (405-414 nm)
Phospholipase A2 (PLA2) catalyzes the hydrolysis of phospholipids at the sn-2 position yielding a free fatty acid and a lysophospholipid. The release of arachidonic acid from membrane phospholipids by PLA2 is believed to be a key step in the control of eicosanoid production within the cell. The Cayman Chemical secretory PLA2 (sPLA2) Assay kit provides an accurate and convenient method for measurement of sPLA2 activity. This assay uses the 1,2-dithio analog of diheptanoyl phosphatidylcholine which serves as a substrate for most PLA2s (for example, bee and cobra venoms, pancreatic, etc.) with the exception of cytosolic PLA2. Upon hydrolysis of the thio ester bond at the sn-2 position by PLA2, free thiols are detected using DTNB (5,5-dithio-bis-(2-nitrobenzoic acid)).
Customers Who Bought This Also Bought