Stationary and Felodipine Baskets, Quality Lab Accessories
Fournisseur: QLA
Certificats
A propos de cet article
QLA manufactures the highest quality Felodipine and Stationary Baskets for use on most dissolution testers.
- Made of high quality 316 Stainless Steel
- All baskets and shafts are made in the USA
- All baskets and shafts are laser marked with a unique serial number
- 40 Mesh baskets are made from sintered mesh
- Modified vessel cover not included
- Custom length shafts and other mesh sizes are available upon request
- 1 cm depth set tool (QLA part# DEPSET-SB) is available
Stationary Baskets and Felodipine Quadrangular Baskets are used with the Paddle Method (APP 2). They are suspended from a special vessel cover (bath specific- see QLA vessel cover page) at a precise distance from the paddle shaft and 1 cm above the paddle. They offer an alternative solution to capsule sinkers for problems associated with floating, sticking and swelling dosage forms. Suitability for certain applications must be verified.
Customers Who Bought This Also Bought