ContactUsLinkComponent
NETosis Assay Kit, Cayman Chemical Company
NETosis Assay Kit, Cayman Chemical Company
# de catalogue 75817-086
Numéro CAS:  
NETosis Assay Kit
NETosis Assay Kit, Cayman Chemical Company
# de catalogue 75817-086
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    75817-086
  • Includes:
    Kit contains: A-23187, Substrate, Reagents, and Plate
  • Supplier No.:
    601010-1
Spécifications
A propos de cet article
For the measurement of NET-derived neutrophil elastase.
  • Induce and detect NET formation in vitro
  • Elastase-based readout
  • Non-dsDNA readout eliminates false positives from DNA
  • Adaptable to multiple species
Cayman's NETosis Assay Kit provides a simple and fast method for studying the process of NETosis ex vivo. Notably, Cayman's NETosis Assay Kit does not depend upon the DNA component of neutrophil extracellular traps (NETs), as DNA release can occur independently of NETosis. In this kit, primary neutrophils are stimulated to release NETs with either PMA or a calcium ionophore (both included). Unbound neutrophil elastase is washed away following NET generation. Following digest of NET DNA by S7 nuclease, the supernatant containing neutrophil elastase is added to a substrate, which is selectively cleaved by elastase to yield a 4-nitroaniline product that adsorbs light at 405 nm. Enough reagents are provided to test 24 sample conditions for NET production, with analysis in duplicate.
Customers Who Bought This Also Bought