Order Entry
ContactUsLinkComponent
cPLA2 Assay Kit, Cayman Chemical Company
cPLA2 Assay Kit, Cayman Chemical Company
# de catalogue 75817-228
Numéro CAS:  
KIT CPLA2 ASSAY PLA2 TYPE IV ASY 96WELLS
cPLA2 Assay Kit, Cayman Chemical Company
# de catalogue 75817-228
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Cat. no.:
    75817-228
  • Includes:
    Kit contains: Substrate, Control, Buffer, Reagents, and 96-Well Plate
  • Supplier No.:
    765021-96

Spécifications

A propos de cet article

For the measurement of cPLA2 activity in purified preparations, cell cultures, or tissue homogenates.

  • Measure cPLA2 activity in cell lysates and tissue homogenates that are known to only contain cPLA2, or in purified enzyme preparations
  • Assay 46 samples in duplicate
  • Measure cPLA2 activity down to 0.0035 U/ml
  • Plate-based colorimetric measurement (405-420 nm)

Phospholipase A2 catalyzes the hydrolysis of fatty acids at the sn-2 position of glycerophospholipids, yielding a free fatty acid and a lysophospholipid as products

Arachidonoyl Thio-PC is a substrate for cPLA2 by virtue of the presence of arachidonic acid at the sn-2 position of the glycerophospholipid. Hydrolysis of the arachidonoyl thioester bond at the sn-2 position by PLA2 releases a free thiol which can be detected by DTNB (5,5'-dithio-bis-(2-nitrobenzoic acid)). This assay can be used to determine the activity of cPLA2 in purified preparations, cell cultures, or tissue homogenates that are known to contain only cPLA2. Use of this assay with preparations containing more than one type of PLA2 will result in the measurement of total PLA2 activity rather than cPLA2 alone. Isozyme-specific cPLA2 activity can be measured by excluding sPLA2 or inhibiting iPLA2 activities in the assay. Each kit contains cPLA2 assay buffer, DTNB/EGTA, Arachidonoyl Thio-PC (substrate), bee venom PLA2 (control), bromoenol lactone solution, a 96 well plate, and complete instructions.

The release of arachidonic acid from membrane phospholipids by these enzymes is believed to be the key step in the biosynthesis of eicosanoids. There are primarily three different kinds of phospholipase A2. They are secretory (sPLA2), calcium-dependent cytosolic (cPLA2), and calcium-independent cytosolic (iPLA2) phospholipase A2. Of these three different types of enzymes, only the cPLA2 exhibits specificity towards arachidonic acid whereas all others can hydrolyze any fatty acid at the sn-2 position.

Customers Who Bought This Also Bought