ContactUsLinkComponent
Electric Whirl
Electric Whirl
# de catalogue 470226-556
Fournisseur:  Wards
Electrical Whirl 95 mm diameter
Electric Whirl
# de catalogue 470226-556
Fournisseur:  Wards
Spécifications
  • Description:
    Electric Whirl
  • Cat. no.:
    470226-556
  • Supplier No.:
    PH0904
Spécifications
A propos de cet article
Demonstrate rotary motion from a charge.
  • For demonstrating the effect of electric discharge from points
The apparatus has a 95 mm diameter, Three-pointed star wheel with all the points curved in one direction. The wheel rotates freely on a pivot point mounted on a short stand and the stand is provided with a 4 mm connection socket.
Customers Who Bought This Also Bought