Order Entry
ContactUsLinkComponent
Human Therapeutic IgG4 ELISA Kit, Cayman Chemical Company
Human Therapeutic IgG4 ELISA Kit, Cayman Chemical Company
# de catalogue 75816-820
Numéro CAS:  
KIT ELISA 96 WELL HUMAN THERAPEUTIC IGG4
Human Therapeutic IgG4 ELISA Kit, Cayman Chemical Company
# de catalogue 75816-820
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Reactivity:
    Human
  • Target protein:
    IgG4
  • Size:
    Kit
  • Detection range:
    7.8 - 500 ng/ml
  • Storage temperature:
    4 °C
  • Cat. no.:
    75816-820
  • Supplier No.:
    500920-96

Spécifications

A propos de cet article

A generic assay for the quantitation of therapeutic human IgG4 antibodies in monkey serum or plasma.

  • Specifically detect human IgG4 antibodies in monkey plasma or serum
  • Not cross reactive to rhesus or cynomolgus immunoglobulins
  • Useful for pharmacokinetic analysis of therapeutic human IgG4 antibodies in monkeys
  • Useful for detecting human IgG4 in mouse or rat plasma or serum

Human therapeutic antibodies have become increasingly common components of early drug discovery and development portfolios in the pharmaceutical and biotech industries. As part of preclinical toxicology assessment of these agents, they are routinely tested in non-human primates, primarily in rhesus or cynomolgus monkeys. In order to assess the pharmacokinetics of the human antibodies in monkey serum, it is necessary to have an assay that is capable of distinguishing the experimentally-introduced human IgG from the endogenous monkey IgG. Cayman’s IgG4 (human) Pharmacokinetic ELISA Kit is an immunometric assay which can be used to measure human IgG4 in monkey or rodent plasma and serum without prior sample purification. The standard curve spans the range of 7.8-500 ng/ml, with an LLOQ of 7.8 ng/ml. The assay has been validated in serum from cynomolgus monkeys.

Delivery: Kit contains: Standard, HRP-Conjugate, Buffers, Reagents, and precoated 96-well Plate
Customers Who Bought This Also Bought