ContactUsLinkComponent
Glucose Uptake Cell-Based Assay Kit, Cayman Chemical Company
Glucose Uptake Cell-Based Assay Kit, Cayman Chemical Company
# de catalogue 75816-992
Numéro CAS:  
KIT GLUCOSE UPTAKE CELL-BASED ASSAY
Glucose Uptake Cell-Based Assay Kit, Cayman Chemical Company
# de catalogue 75816-992
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    75816-992
  • Includes:
    Kit contains: 2-NBDG, Apigenin, Propidium Iodide solution, and Buffer tablet
  • Supplier No.:
    600470-1
Spécifications
A propos de cet article
A cell-based assay for studying modulators of cellular glucose uptake.
  • Measure glucose uptake by cultured cells
  • Employs 2-NBDG, a fluorescent deoxyglucose analog
  • An inhibitor of glucose transport is included as a control
  • Convenient tool for studying modulators of glucose uptake
Glucose metabolism is a primary source of energy and biomaterials for the maintenance of cell homeostasis. Extra glucose is stored in the muscles and liver as glycogen which is hydrolyzed to glucose and released into the blood when needed. The rate of glucose uptake in cells is dynamic and tightly regulated by hormones and/or growth factors including insulin. Cancer cells exhibit increased glucose uptake and metabolism by aerobic glycolysis in order to support a high rate of proliferation. Chemicals that block glucose uptake by cancer cells have been shown to have anti-cancer effects. Cayman’s Glucose Uptake Cell-based Assay Kit provides a convenient tool for studying modulators of cellular glucose uptake. The kit employs 2-NBDG, a fluorescently-labeled deoxyglucose analog, as a probe for the detection of glucose taken up by cultured cells. Apigenin, a flavonoid that has been reported to be an inhibitor of glucose transport, is included as a control.
Customers Who Bought This Also Bought