ContactUsLinkComponent
Glycolysis Cell-Based Assay Kit, Cayman Chemical Company
Glycolysis Cell-Based Assay Kit, Cayman Chemical Company
# de catalogue 75816-990
Numéro CAS:  
KIT ASSAY GLYCOLYSIS CELL-BASED 156 UM
Glycolysis Cell-Based Assay Kit, Cayman Chemical Company
# de catalogue 75816-990
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    75816-990
  • Includes:
    Kit contains: Standard, Buffer tablet, Substrate, Enzyme mixture, and Cofactor
  • Supplier No.:
    600450-1
Spécifications
A propos de cet article
A colorimetric-based method for detecting L-lactate in culture medium
  • Detect L-lactate, the end product of glycolysis
  • Can be adapted to high-throughput screening
  • Measure L-lactate in cell culture supernatant down to 156 µM
Cayman’s Glycolysis Cell-Based Assay Kit provides a colorimetric method for detecting L-lactate, the end product of glycolysis, produced and secreted by cultured cells. In this assay, lactate dehydrogenase catalyzes the reaction between NAD+ and lactate, yielding pyruvate and NADH. The NADH directly reduces a tetrazolium salt (INT) to a colored formazan which absorbs between 490 and 520 nm. The quantity of formazan produced is proportional to the quantity of lactate in the culture medium, and is thus an indirect measurement of glycolysis. This assay can be adapted to high-throughput screening to examine glycolytic regulators in a cell-based system. In addition, the non-invasive sampling leaves the cells intact and thus allows multiplex testing for additional markers.
Customers Who Bought This Also Bought