ContactUsLinkComponent
Triglyceride Colorimetric Assay Kit, Cayman Chemical
Triglyceride Colorimetric Assay Kit, Cayman Chemical
# de catalogue 75816-756
Numéro CAS:  
KIT ASSAY TRIGLYCERIDE COLORIMETRIC 96WL
Triglyceride Colorimetric Assay Kit, Cayman Chemical
# de catalogue 75816-756
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    75816-756
  • Range:
    3.125-200 mg/dl
  • Includes:
    Kit contains: Standard, Buffer, Reagents, and 96-well plate
  • Supplier No.:
    10010303-96
Spécifications
A propos de cet article
A simple, reproducible, and sensitive tool for assaying triglycerides in plasma, serum, cell lysates, and tissue homogenate samples.
  • Measure TG levels in plasma, serum, cell lysates, and tissue homogenates
  • Assay 26 samples in triplicate and 40 samples in duplicate
  • Assay Range: 3.125-200 mg/dl
  • Incubation: 15 minutes | Read: Colorimetric at 530-550 nm
The measurement of triglyceride levels, in conjunction with other lipid assays, are useful in the diagnosis of primary and secondary hyperlipoproteinemia, dyslipidemia, and triglyceridemia. Cayman’s Triglyceride Assay Kit provides a simple, reproducible, and sensitive tool for assaying triglycerides in plasma, serum, cell lysates, and tissue homogenate samples. The assay is initiated with the enzymatic hydrolysis of the triglycerides by lipase to produce glycerol and free fatty acids. The glycerol released is subsequently measured by a coupled enzymatic reaction system with a colorimetric readout at 540 nm.
Customers Who Bought This Also Bought