ContactUsLinkComponent
Glucose Colorimetric Assay Kit, Cayman Chemical Company
Glucose Colorimetric Assay Kit, Cayman Chemical Company
# de catalogue 75817-348
Numéro CAS:  
KIT GLUCOSE COLORIMETRIC ASSAY 192 WELLS
Glucose Colorimetric Assay Kit, Cayman Chemical Company
# de catalogue 75817-348
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    75817-348
  • Range:
    2.5-25 mg/dl
  • Includes:
    Kit contains: Standard, Buffer, Reagents, and 96-well plate
  • Supplier No.:
    10009582-192
Spécifications
A propos de cet article
For analysis of glucose in plasma, serum, and urine.
  • Measure glucose in plasma, serum, and urine
  • Assay 40 samples in duplicate
  • Assay Range: 2.5-25 mg/dl
  • Plate-based colorimetric measurement (500-520 nm)
Glucose, a monosaccharide (or simple sugar), is the most important carbohydrate in biology. Transported via the blood stream, it is the primary source of energy for the body’s cells. Glucose level is tightly regulated in the human body. Failure to maintain blood glucose in the normal range leads to conditions of persistently high (hyperglycemia) or low (hypoglycemia) blood sugar. Diabetes mellitus, characterized by persistent hyperglycemia, is the most prominent disease related to failure of blood sugar regulation. Cayman’s Glucose Colorimetric Assay Kit provides a simple, reproducible, and sensitive tool for assaying glucose in plasma, serum, and urine. The glucose assay uses the glucose oxidase-peroxide reaction for the determination of glucose concentrations. In this assay, glucose is oxidized to δ-gluconolactone with concomitant reduction of the Flavin adenine dinucleotide (FAD)-dependent enzyme glucose oxidase. The reduced form of glucose oxidase is regenerated to its oxidized form by molecular oxygen to produce hydrogen peroxide. Finally, with horseradish peroxidase as a catalyst, hydrogen peroxide reacts with 3,5-dichloro-2-hydroxybenzenesulfonic acid and 4-aminoantipyrine to generate a pink dye with an optimal absorption at 514 nm.
Customers Who Bought This Also Bought