Wood Cane
# de catalogue 470226-524
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Description:Wood Cane
- Cat. no.:470226-524
- Supplier No.:MDS80212
Spécifications
A propos de cet article
Wooden canes provide an attractive, classic style for users who desire a traditional walking aid.
- For Patients Who Need Additional Assistance with Balance
- Weight Capacity: 136 kg (300 lb.)
- Traditional Walking Aid
These canes are designed for patients who need additional assistance with balance. They have a weight capacity of 136 kg (300 lb.).
The standard handle lets users hook the cane over their arm while opening doors or performing other tasks. The canes cannot adjust to height.
Customers Who Bought This Also Bought