Exam Table Paper
# de catalogue 470226-510
Fournisseur: Avantor
Spécifications
- Description:Smooth exam table paper
- Dimensions:18"×225 ft.
- Supplier No.:BXT62085520
Spécifications
A propos de cet article
High-quality white papers feature either a crepe or smooth finish.
- Durable
- Comfortable
- Crepe or Smooth Finish
- Lightweight
- White
Papers are lightweight and comfortable for the patient.
They're also durable to help protect the examination table from moisture, soil, or contamination.
Packaging: Packaged 12 rolls of paper per case.
Customers Who Bought This Also Bought