A propos de cet article
Cincinnati Surgical products, made from high grade British surgical steel, hold an excellent reputation for sharpness and consistency in size, shape, and durability
- 200% visual inspection
- Packaged with VCI paper to protect the blade from corrosion
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
- Standard range
- White handle
They have been a hallmark of this brand for decades.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 75870-488
- 75870-622
- 75870-624
- 75870-480
- 75870-482
- 75870-484
- 75870-486
- 75870-640
- 75870-490
- Environmental certification:
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001