TLC Plates, Silica Gel 60, Unmodified Layers, Polyester Sheets, SIL G, POLYGRAM®, Macherey-Nagel
Fournisseur: Macherey-Nagel
Certificats
A propos de cet article
Ready-to-use sheets
- Transparent
- Unbreakable but easy to cut
- Chemical resistant against acids and bases
- Returns to planarity after bending
- Temperature stable up to 185 °C
The binder is stable in almost all organic solvents and resistant towards aggressive visualisation reagents.
Customers Who Bought This Also Bought