O-Cedar® Rinse Fresh® Iron Scrub Brush, Vileda Professional
# de catalogue 75876-112
Fournisseur: Vileda Professional - FHP
Spécifications
- Bristle Color:Blue
- Handle Material:Plastic
- Description:Rinse Fresh® Iron Scrub Brush
- Total length:15.2 cm (6")
- Cat. no.:75876-112
- Handle Color:White/Red
- Supplier No.:148187
Spécifications
A propos de cet article
This brush easily scrub away tough stains, mildew, soap scum and grime.
- Ergonomic, sturdy handle is rubberized for comfort and better grip when wet
- Rinse Fresh® channels allow for easy, effective rinsing so brush stays clean and fresh
- Designed to keep hands drier and protect your knuckles when scrubbing
- Antibacterial protection built into the bristles prevents odor-causing bacteria
- Bristles have excellent bend recovery and resist stains
- Replace every 90 days
Designed to keep hands drier and protect your knuckles when scrubbing, this multi-purpose brush features an angled head for those hard-to-reach corners.
Customers Who Bought This Also Bought