Laboratory Dry Sterilizer, Yamato
Fournisseur: Yamato Scientific
Certificats
A propos de cet article
The low-cost, programmable laboratory dry sterilizer is easy to operate.
- Two 30 mm dia. exhaust ports on top
- Maximum temperature reaching time: ~70 min
- Two shelf plates and four brackets included
- Natural convection
- Iron chromium wire heater
The sterilizer features a stainless steel interior and an exterior with a chrome-free, electro-galvanized carbon steel sheet coated with a chemical-proof, baked-on finish.
A green LED display indicates the measured temperature, and a red LED display indicates the setting temperature. An overheat prevention device and an earth leakage breaker are provided. Self-diagnostic functions ensure safe operation.
It's equipped with calibration offset, a lock function, and power-failure recovery mode. The natural convection oven has a temperature accuracy of 20 °C (at 260 °C).
Caution: High Temperature Components. Never use explosive or flammable material. Never use in flammable or explosive gas atmosphere.
Spécifications
- Catalog No:
- 76550-284
- 76550-282
- 76550-280
- 76550-286
- Control type:
- PID Temperature Control
- PID Temperature Control
- PID Temperature Control
- PID Temperature Control
- Max. load per shelf:
- ~15 kg/pc
- ~15 kg/pc
- ~15 kg/pc
- ~15 kg/pc
- Programs:
- 6 Pattern: 30 steps × 1, 15 steps × 2, and 10 steps × 3 (pattern repeat function included)
- 6 Pattern: 30 steps × 1, 15 steps × 2, and 10 steps × 3 (pattern repeat function included)
- 6 Pattern: 30 steps × 1, 15 steps × 2, and 10 steps × 3 (pattern repeat function included)
- 6 Pattern: 30 steps × 1, 15 steps × 2, and 10 steps × 3 (pattern repeat function included)
- Sensor:
- K Thermocouple
- K Thermocouple
- K Thermocouple
- K Thermocouple
- Timer:
- 1 min - 99 hrs, 59 min and 100 - 999 hrs, 30 min (with timer wait function)
- 1 min - 99 hrs, 59 min and 100 - 999 hrs, 30 min (with timer wait function)
- 1 min - 99 hrs, 59 min and 100 - 999 hrs, 30 min (with timer wait function)
- 1 min - 99 hrs, 59 min and 100 - 999 hrs, 30 min (with timer wait function)