A propos de cet article
The air-jacket technology of these incubators ensures even and efficient heat distribution throughout the chamber.
- Natural Convection System
- Operating Temperature Range: Room Temperature +5 to ~80°C
- Heater Control Circuit: SSR Drive System
- Temperature Accuracy: ±1.0 °C
- Temperature Control Accuracy: ±0.5 °C (at 37 °C)
- Power Outage Compensation
- Digital PID Controller
The space-saving incubators have a stainless steel interior and a cold-rolled steel plate exterior with a melamine-resin baking finish
The digital setting display and timer are easy to use. A green LED display indicates the measurement temperature, and a red LED display indicates the setting temperature. Both temperatures can be set with keys. Operation functions include fixed temperature, auto start and stop, calibration offset, key lock, and quick auto stop. Safety features include temperature sensor and measurement temperature error and automatic overheat prevention. Self-diagnostic functions make operation safe and error recovery fast.
Benchtop incubators feature stainless steel heating pipes, and larger incubators feature iron-chrome wire heaters. Shelves are provided.
Spécifications
- Catalog No:
- 75846-604
- 75846-602
- 75846-606
- 75846-718
- 75846-720
- 75846-702
- 75846-704
- 75846-600
- 75846-706
- 75846-616
- 75846-708
- 75846-710
- 75846-612
- 75846-712
- 75846-610
- 75846-714
- 75846-608
- Max. load per shelf:
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- ~15 kg
- Timer:
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- 1 Minute to 99 Hours, 59 Minutes and 100 to 999 Hours, 50 Minutes (including timer waiting function)
- Sensor:
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- K Thermocouple Sensor
- Insulation:
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator
- Glass Fiber Heat Insulator