Programmable Natural Convection Oven, Yamato | Avantor
Programmable Natural Convection Oven, Yamato
Programmable Natural Convection Oven, Yamato
Supplier:  
Certificates
Programmable Natural Convection Oven, Yamato
Programmable Natural Convection Oven, Yamato
0 Options Available Below
Product Details & Documents
These constant-temperature ovens feature easy-to-use program settings.
  • Six-Pattern PID Program Controller
  • Simultaneous Display of Set Constant and Measured Temperature
  • Cable Hole: 30 mm dia. on Right Side
  • Exhaust Port: Two 30 mm dia. on Top
  • Two Stainless Steel Plates with Four Brackets Provided
They take approximately 90 minutes (room temperature: ±5 to 260 °C) to heat
The interior material is stainless steel, and the exterior is made of a cold-rolled steel plate with melamine resin baking finish. The glass observation window is chemically strengthened. A SSR control adjusts the heater circuit. Operation functions include fixed-temperature operation, quick auto stop, and auto stop and start. A calibration offset function, a key lock function, uninterruptible power for memory, and a pattern-repeat function are also included. Safety features such as self-diagnostic functions, overheat prevention, and electric leakage breaker with over-current protection are provided as well.
The temperature setting can be digitally adjusted. Green LED lights display measurement temperatures, and red LED lights display setting temperatures.
Informations sur les accessoires: Stainless steel shelf available separately.
Caution: Never use in flammable or explosive gas atmosphere. Never use explosive or flammable material. High-temperature components
Specifications for Product Family
Spécifications
  • Catalog No:
  • 75846-332
  • 75846-334
  • 75846-336
  • 75846-338
  • Control type:
  • Six-Pattern PID Program Controller
  • Six-Pattern PID Program Controller
  • Six-Pattern PID Program Controller
  • Six-Pattern PID Program Controller
  • Insulation:
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Sensor:
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • Timer:
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes