Dissolution Baskets and Basket Accessories for Apparatus 1
Fournisseur: QLA
Certificats
A propos de cet article
QLA is the only on-site basket manufacturer in the industry.
- Made of high quality 316 Stainless Steel (excludes Suppository Baskets)
- All baskets made in the USA
- Laser marked with unique serial number (excludes Suppository baskets)
- 40 Mesh baskets are made from sintered mesh
- Variety of mesh sizes available
QLA offers both clip style and O-ring Push-on style baskets for most dissolution baths.
Our 40 mesh baskets are made with sintered 316 Stainless Steel. The sintering process essentially creates a welded joint at each overlapping wire junction. Sintered baskets are stronger and last longer. They retain original dimensions and geometry and resist wobble. Most conventional baskets are not sintered and weaken over time. Critical basket geometry is verified on an optical comparator.
Certifications: All baskets are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an extra cost.
Customers Who Bought This Also Bought