KIMBLE® PTFE-Faced Red Rubber Septa, DWK Lifesciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Highly chemical-resistant PTFE facing maintains integrity of contents.
- Economical and excellent resealability
- Low extractables
- Resists coring
- PTFE thickness: 0.002"
Customers Who Bought This Also Bought