ContactUsLinkComponent
PiBlue™ Pho Phosphate Assay Kit, BioAssay Systems
PiBlue™ Pho Phosphate Assay Kit, BioAssay Systems
# de catalogue 75878-188
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
PiBlue* Phosphate Assay Kit 500tests
PiBlue™ Pho Phosphate Assay Kit, BioAssay Systems
# de catalogue 75878-188
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Phosphate Assay Kit
  • Size:
    500 assays
  • Method:
    OD620 nm
  • Sample:
    Enzyme reactions with free phosphate releases.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-188
  • Supplier No.:
    POPB-500
Spécifications
A propos de cet article
For sensitive and high-throughput phosphate determination.
  • Very stable reagent
  • High sensitivity and wide detection range (0.4 - 50 μM phosphate)
  • Can be readily automated on HTS liquid handling systems
The PiBlue™ Phosphate Assay Kit is based on a proprietary formulation of the malachite green dye. The PiBlue™ reagent forms a blue colored complex with free orthophosphate. The rapid color formation from the reaction can be conveniently measured on a spectrophotometer (600 - 660 nm) or on a plate reader. The non-radioactive colorimetric assay kits have been optimized to offer superior sensitivity and prolonged. Shelf Life. The assay is simple and fast, involving a single addition step for phosphate determination. Assays can be performed in tubes, cuvettes or multi-well plates. The assays can be conveniently executed in 96-well plates for high-throughput screening of enzyme inhibitors.
Customers Who Bought This Also Bought