ContactUsLinkComponent
EnzyChrom™ Xanthine Assay Kit, BioAssay Systems
EnzyChrom™ Xanthine Assay Kit, BioAssay Systems
# de catalogue 75878-178
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Xanthine Assay Kit 100 tests
EnzyChrom™ Xanthine Assay Kit, BioAssay Systems
# de catalogue 75878-178
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Xanthine Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, FL530/585 nm
  • Sample:
    Cell lysate, serum, and other biological samples.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-178
  • Supplier No.:
    EXAN-100
Spécifications
A propos de cet article
For quantitative determination of xanthine and drug effects on its metabolism.
  • Linear detection range 0.01 - 2.5 mM xanthine for colorimetric assays and 3 - 250 µM for fluorimetric assays
  • Fast and high-throughput. Can be automated to test thousands of samples per day
Xanthine is a purine base that can be found in most animal tissues and fluids
BioAssay Systems’ xanthine assay kit uses a single Working Reagent that combines the xanthine oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to xanthine concentration in the sample.
It is a product in the purine degradation pathway, produced by guanine deaminase from guanine, and by xanthine oxidoreductase from hypoxanthine. Xanthine is degraded to uric acid by xanthine oxidase. Clinically, xanthine and its derivatives act on sleep-inducing adenosine receptors as antagonists. Simple, direct and high-throughput assays for measuring xanthine find wide applications in research and drug discovery.
Customers Who Bought This Also Bought